siRNA Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022158.3 | NP_071441.1 | 3 | 422 | ||
| GenBank mRNA | AJ404615.1 | - | 3 | 382 | ||
| AK054838.1 | - | 3 | 1372 | |||
| AK294871.1 | - | 3 | 383 | |||
| BC042680.1 | - | 3 | 366 | 5195472 | ||

FN3K
Genomic Map