siRNA Details
| Gene Symbol: CCDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130917.1 | 4 | 641 | |||
| NR_130918.1 | 3 | 418 | ||||
| NR_130919.1 | 4 | 572 | ||||
| NR_130920.1 | 4 | 589 | ||||
| GenBank mRNA | BC070152.1 | - | 4 | 641 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC26| Gene Symbol: CCDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130917.1 | 4 | 641 | |||
| NR_130918.1 | 3 | 418 | ||||
| NR_130919.1 | 4 | 572 | ||||
| NR_130920.1 | 4 | 589 | ||||
| GenBank mRNA | BC070152.1 | - | 4 | 641 | ||