siRNA Details
| Gene Symbol: SSMEM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145268.3 | NP_660311.1 | 3 | 662 | ||
| XM_005250141.3 | XP_005250198.1 | 3 | 569 | |||
| XM_011515794.2 | XP_011514096.1 | 3 | 526 | |||
| XM_011515795.2 | XP_011514097.1 | 3 | 519 | |||
| GenBank mRNA | AK097635.1 | - | 3 | 648 | ||
| BC017587.2 | - | 3 | 666 | 4838989 | ||

SSMEM1
Genomic Map