siRNA Details
| Gene Symbol: FAM185A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145268.1 | NP_001138740.1 | 8 | 2194 | ||
| NM_001145269.1 | NP_001138741.1 | 7 | 1843 | |||
| NR_026879.1 | 8 | 1953 | ||||
| GenBank mRNA | AK054745.1 | - | 8 | 1863 | ||
| BC029175.1 | - | 8 | 2153 | |||

FAM185A
Genomic Map