siRNA Details
| Gene Symbol: TMEM155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317837.1 | NP_001304766.1 | 6 | 1302 | ||
| NM_001317838.1 | NP_001304767.1 | 6 | 1299 | |||
| NM_001317839.1 | NP_001304768.1 | 6 | 1177 | |||
| NM_001317841.1 | NP_001304770.1 | 5 | 1592 | |||
| NM_001317842.1 | NP_001304771.1 | 5 | 1436 | |||
| NM_152399.3 | NP_689612.3 | 6 | 1458 | |||
| XM_005262737.2 | XP_005262794.1 | 6 | 1266 | |||
| XM_017007725.1 | XP_016863214.1 | 6 | 1172 | |||
| XM_017007726.1 | XP_016863215.1 | 5 | 1112 | |||
| GenBank mRNA | AK055396.1 | - | 6 | 1179 | ||
| BC047657.1 | - | 6 | 1458 | 5264721 | ||

TMEM155
Genomic Map