siRNA Details
| Gene Symbol: JMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152405.4 | NP_689618.4 | 2 | 1687 | ||
| XM_005248430.2 | XP_005248487.1 | 2 | 1687 | |||
| XM_011543155.2 | XP_011541457.1 | 2 | 1687 | |||
| GenBank mRNA | AK095189.1 | - | 1 | 88 | ||
| BC130624.1 | - | 1 | 113 | 40146655 | ||
| CN372786.1 | - | 2 | 327 | |||
| DW425980.1 | - | 1 | 71 | |||

JMY
Genomic Map