siRNA Details
| Gene Symbol: C5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135586.1 | NP_001129058.1 | 2 | 1501 | ||
| NM_152409.3 | NP_689622.2 | 2 | 1314 | |||
| XM_005271889.3 | XP_005271946.1 | 2 | 2150 | |||
| XM_017009049.1 | XP_016864538.1 | 2 | 1669 | |||
| XM_017009050.1 | XP_016864539.1 | 2 | 1277 | |||
| GenBank mRNA | AK094881.1 | - | 2 | 1451 | ||
| BC053677.1 | - | 2 | 1314 | 6166677 | ||
| BX648529.1 | - | 2 | 1381 | |||

C5orf24
Genomic Map