siRNA Details
| Gene Symbol: VPS37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145152.1 | NP_001138624.1 | 4 | 789 | ||
| NM_152415.2 | NP_689628.2 | 5 | 864 | |||
| XM_005273400.2 | XP_005273457.1 | 6 | 874 | |||
| XM_005273401.2 | XP_005273458.1 | 5 | 820 | |||
| XM_006716286.1 | XP_006716349.1 | 5 | 829 | |||
| XM_017013021.1 | XP_016868510.1 | 5 | 829 | |||
| XM_017013022.1 | XP_016868511.1 | 5 | 829 | |||
| XM_017013023.1 | XP_016868512.1 | 6 | 874 | |||
| XM_017013024.1 | XP_016868513.1 | 5 | 820 | |||
| XM_017013025.1 | XP_016868514.1 | 6 | 874 | |||
| GenBank mRNA | AF547097.1 | - | 4 | 558 | ||
| AK057204.1 | - | 5 | 785 | |||
| AY033079.2 | - | 5 | 662 | |||
| BC022363.1 | - | 1 | 20 | |||
| BC067754.1 | - | 5 | 777 | 5275060 | ||

VPS37A
Genomic Map