siRNA Details
| Gene Symbol: FAM69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152421.3 | NP_689634.2 | 5 | 1021 | ||
| XM_006716954.3 | XP_006717017.1 | 5 | 1026 | |||
| XM_011518238.2 | XP_011516540.1 | 6 | 1152 | |||
| XM_017014273.1 | XP_016869762.1 | 4 | 1059 | |||
| GenBank mRNA | AF318355.1 | - | 3 | 1766 | ||
| AJ420457.1 | - | 4 | 861 | |||
| BC032097.2 | - | 5 | 1015 | 3161575 | ||

FAM69B
Genomic Map