siRNA Details
| Gene Symbol: SRFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152546.2 | NP_689759.2 | 8 | 1913 | ||
| GenBank mRNA | AK058015.1 | - | 8 | 1914 | ||
| BC017102.1 | - | 3 | 1496 | |||
| BC031222.1 | - | 8 | 1884 | 5297296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRFBP1| Gene Symbol: SRFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152546.2 | NP_689759.2 | 8 | 1913 | ||
| GenBank mRNA | AK058015.1 | - | 8 | 1914 | ||
| BC017102.1 | - | 3 | 1496 | |||
| BC031222.1 | - | 8 | 1884 | 5297296 | ||