siRNA Details
| Gene Symbol: BMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152556.2 | NP_689769.2 | 2 | 415 | ||
| GenBank mRNA | AK056380.1 | - | 2 | 415 | ||
| AK299901.1 | - | 4 | 261 | |||
| AL834437.1 | - | 2 | 210 | |||
| BC114615.1 | - | 2 | 407 | 40032372 | ||
| DA738444.1 | - | 2 | 415 | |||

BMT2
Genomic Map