siRNA Details
Gene Symbol: BMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152556.2 | NP_689769.2 | 5 | 2151 | ||
GenBank mRNA | AK056380.1 | - | 5 | 2152 | ||
AL834437.1 | - | 5 | 1946 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152556.2 | NP_689769.2 | 5 | 2151 | ||
GenBank mRNA | AK056380.1 | - | 5 | 2152 | ||
AL834437.1 | - | 5 | 1946 |