siRNA Details
| Gene Symbol: C9orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152571.2 | NP_689784.1 | 1 | 1946 | ||
| GenBank mRNA | AK055336.1 | - | 1 | 1898 | ||
| AK094098.1 | - | 1 | 1217 | |||
| BC036221.1 | - | 1 | 1946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf163| Gene Symbol: C9orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152571.2 | NP_689784.1 | 1 | 1946 | ||
| GenBank mRNA | AK055336.1 | - | 1 | 1898 | ||
| AK094098.1 | - | 1 | 1217 | |||
| BC036221.1 | - | 1 | 1946 | |||