siRNA Details
| Gene Symbol: TMEM192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100389.1 | NP_001093859.1 | 3 | 476 | ||
| XM_011531717.2 | XP_011530019.1 | 3 | 517 | |||
| XM_011531718.2 | XP_011530020.1 | 3 | 486 | |||
| GenBank mRNA | AK095801.1 | - | 4 | 548 | ||
| BC036301.1 | - | 3 | 421 | 4778254 | ||
| BX538029.1 | - | 3 | 434 | |||
| BX648915.1 | - | 3 | 432 | |||

TMEM192
Genomic Map