siRNA Details
| Gene Symbol: RWDD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307922.1 | NP_001294851.1 | 3, 4 | 483 | ||
| NM_152682.3 | NP_689895.2 | 3, 4 | 487 | |||
| GenBank mRNA | AK021702.1 | - | 3, 4 | 411 | ||
| AK315515.1 | - | 3, 4 | 421 | |||
| AL540937.3 | - | 3, 4 | 424 | |||
| AL559547.3 | - | 3, 4 | 420 | |||
| BC017472.1 | - | 3, 4 | 422 | 3909581 | ||
| BC067752.1 | - | 3, 4 | 446 | 5270461 | ||
| BC107432.1 | - | 3, 4 | 268 | 6104330 | ||
| DA766837.1 | - | 3, 4 | 486 | |||

RWDD4
Genomic Map