siRNA Details
| Gene Symbol: TMEM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097620.1 | NP_001091089.1 | 9 | 4456 | ||
| XM_017011815.1 | XP_016867304.1 | 8 | 5448 | |||
| GenBank mRNA | AK096817.1 | - | 2 | 1983 | ||
| AK126312.1 | - | 8 | 4256 | |||

TMEM184A
Genomic Map