siRNA Details
| Gene Symbol: C7orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152706.3 | NP_689919.1 | 2 | 773 | ||
| XM_011515904.2 | XP_011514206.1 | 2 | 2990 | |||
| GenBank mRNA | BC028365.2 | - | 2 | 773 | 4838806 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf62| Gene Symbol: C7orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152706.3 | NP_689919.1 | 2 | 773 | ||
| XM_011515904.2 | XP_011514206.1 | 2 | 2990 | |||
| GenBank mRNA | BC028365.2 | - | 2 | 773 | 4838806 | |