siRNA Details
| Gene Symbol: ATXN7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152749.3 | NP_689962.1 | 4 | 464 | ||
| XM_011515954.2 | XP_011514256.1 | 5 | 547 | |||
| GenBank mRNA | AK095475.1 | - | 4 | 378 | ||
| BC030616.2 | - | 4 | 461 | 4796569 | ||

ATXN7L1
Genomic Map