siRNA Details
| Gene Symbol: MTURN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152793.2 | NP_690006.2 | 3 | 1669 | ||
| GenBank mRNA | AL834391.1 | - | 3 | 1606 | ||
| AY054121.1 | - | 3 | 1465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTURN| Gene Symbol: MTURN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152793.2 | NP_690006.2 | 3 | 1669 | ||
| GenBank mRNA | AL834391.1 | - | 3 | 1606 | ||
| AY054121.1 | - | 3 | 1465 | |||