siRNA Details
| Gene Symbol: TMEM130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134450.1 | NP_001127922.1 | 3 | 605 | ||
| NM_001134451.1 | NP_001127923.1 | 2 | 299 | |||
| NM_152913.2 | NP_690877.1 | 3 | 605 | |||
| GenBank mRNA | AK124634.1 | - | 3 | 1482 | ||
| AK225783.1 | - | 3 | 548 | |||
| AK294658.1 | - | 3 | 421 | |||
| AK295543.1 | - | 2 | 263 | |||
| AK312364.1 | - | 3 | 475 | |||
| AL832523.1 | - | 2 | 339 | |||
| AL834348.1 | - | 3 | 451 | |||
| AL834349.1 | - | 3 | 421 | |||
| AY358583.1 | - | 4 | 537 | |||
| BC030793.1 | - | 3 | 574 | 5263130 | ||
| BC037895.1 | - | 2 | 211 | 5275566 | ||
| DA154119.1 | - | 2 | 318 | |||

TMEM130
Genomic Map