siRNA Details
| Gene Symbol: TMEM74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153015.2 | NP_694560.1 | 2 | 961 | ||
| GenBank mRNA | AK055230.1 | - | 2 | 887 | ||
| BC030710.2 | - | 2 | 961 | 4819093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM74| Gene Symbol: TMEM74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153015.2 | NP_694560.1 | 2 | 961 | ||
| GenBank mRNA | AK055230.1 | - | 2 | 887 | ||
| BC030710.2 | - | 2 | 961 | 4819093 | ||