siRNA Details
Gene Symbol: C6orf223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171992.2 | NP_001165463.1 | 4 | 1650 | ||
NM_001324369.1 | NP_001311298.1 | 3 | 1480 | |||
NM_153246.5 | NP_694978.2 | 4 | 1607 | |||
XM_017010425.1 | XP_016865914.1 | 4 | 2689 | |||
GenBank mRNA | BC029071.2 | - | 4 | 1617 | 4341806 | |
BC032706.1 | - | 3 | 1921 | 5497384 |
