siRNA Details
| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184767.1 | NP_001171696.1 | 5 | 816 | ||
| NM_153346.4 | NP_699177.2 | 5 | 816 | |||
| XM_011545447.1 | XP_011543749.1 | 5 | 798 | |||
| XM_017029266.1 | XP_016884755.1 | 5 | 798 | |||
| XM_017029267.1 | XP_016884756.1 | 5 | 798 | |||
| GenBank mRNA | AK128155.1 | - | 5 | 798 | ||
| BC037301.2 | - | 5 | 796 | 4827886 | ||
| BC096745.1 | - | 5 | 749 | 4830576 | ||

BEND2
Genomic Map