siRNA Details
| Gene Symbol: TMEM67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142301.1 | NP_001135773.1 | 20 | 1891 | ||
| NM_153704.5 | NP_714915.3 | 19 | 1960 | |||
| NR_024522.1 | 19 | 1960 | ||||
| XM_006716686.3 | XP_006716749.1 | 18 | 1798 | |||
| XM_011517363.2 | XP_011515665.1 | 10 | 1050 | |||
| GenBank mRNA | AK026742.1 | - | 14 | 1266 | ||
| AK092244.1 | - | 20 | 1848 | |||
| AK094935.1 | - | 19 | 1884 | |||
| AK297676.1 | - | 4 | 613 | |||
| AK308735.1 | - | 10 | 1027 | |||
| BC032835.2 | - | 19 | 1878 | 4825770 | ||
| BC054338.1 | - | 19 | 1887 | |||
| BX648768.1 | - | 19 | 1904 | |||
| KU178827.1 | - | 19 | 1859 | |||
| KU178828.1 | - | 18 | 1705 | |||

TMEM67
Genomic Map