siRNA Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131065.1 | NP_001124537.1 | 6 | 1778 | ||
| NM_001131066.1 | NP_001124538.1 | 6 | 623 | |||
| NM_173362.3 | NP_775498.1 | 5 | 1680 | |||
| XM_011543360.2 | XP_011541662.1 | 7 | 1666 | |||
| XM_011543361.2 | XP_011541663.1 | 5 | 558 | |||
| XM_017009413.1 | XP_016864902.1 | 6 | 790 | |||
| GenBank mRNA | BC024023.1 | - | 4 | 397 | 4670524 | |
| BC035110.2 | - | 5 | 1678 | 5262922 | ||
| BC048979.1 | - | 5 | 1705 | 4838006 | ||
| BI551847.1 | - | 6 | 626 | |||
| BM263054.1 | - | 5 | 587 | |||

RFESD
Genomic Map