siRNA Details
| Gene Symbol: C4orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099783.1 | NP_001093253.1 | 6 | 670 | ||
| NM_173487.2 | NP_775758.2 | 6 | 942 | |||
| XM_005262734.4 | XP_005262791.1 | 6 | 937 | |||
| GenBank mRNA | AK091022.1 | - | 6 | 931 | ||
| BC016358.2 | - | 2 | 283 | |||
| BC032582.1 | - | 6 | 873 | 5494700 | ||
| BX538164.1 | - | 6 | 883 | |||

C4orf33
Genomic Map