siRNA Details
| Gene Symbol: MROH2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173489.4 | NP_775760.3 | 25 | 2955 | ||
| XM_011513952.1 | XP_011512254.1 | 25 | 2941 | |||
| XM_011513953.1 | XP_011512255.1 | 24 | 2879 | |||
| GenBank mRNA | AK126005.1 | - | 10 | 824 | ||
| AK128183.1 | - | 15 | 1421 | |||
| AL832763.1 | - | 20 | 1880 | |||
| BC044937.1 | - | 13 | 1427 | |||
| BC052962.1 | - | 25 | 2916 | 4838071 | ||
| CR749346.1 | - | 20 | 1856 | |||

MROH2B
Genomic Map