siRNA Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 4, 5 | 773 | ||
| XM_005262938.3 | XP_005262995.1 | 4, 5 | 1510 | |||
| XM_005262939.3 | XP_005262996.1 | 3, 4 | 575 | |||
| XM_005262940.4 | XP_005262997.1 | 2, 3 | 712 | |||
| XM_011531881.2 | XP_011530183.1 | 4, 5 | 857 | |||
| XM_011531882.2 | XP_011530184.1 | 4, 5 | 1521 | |||
| GenBank mRNA | BC034385.1 | - | 4, 5 | 774 | 5180678 | |

RNF175
Genomic Map