siRNA Details
| Gene Symbol: DTWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 629 | ||
| NM_173666.3 | NP_775937.1 | 6 | 850 | |||
| XM_011543338.2 | XP_011541640.2 | 7 | 2243 | |||
| XM_011543340.2 | XP_011541642.1 | 7 | 742 | |||
| GenBank mRNA | AK091296.1 | - | 6 | 790 | ||
| BC114508.1 | - | 6 | 786 | 40024023 | ||
| BC114955.1 | - | 6 | 786 | 40024022 | ||
| BX648369.1 | - | 6 | 629 | |||

DTWD2
Genomic Map