siRNA Details
| Gene Symbol: LOC100996634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277339.1 | NP_001264268.1 | 2 | 296 | ||
| XM_017010153.1 | XP_016865642.1 | 4 | 4593 | |||
| XM_017010154.1 | XP_016865643.1 | 4 | 4593 | |||
| GenBank mRNA | AK094715.1 | - | 2 | 296 | ||

LOC100996634
Genomic Map