siRNA Details
| Gene Symbol: C8orf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173687.3 | NP_775958.1 | 6 | 1034 | ||
| NR_126398.1 | 8 | 1216 | ||||
| NR_126399.1 | 6 | 1025 | ||||
| GenBank mRNA | AK094450.1 | - | 6 | 982 | ||
| BC063497.1 | - | 6 | 936 | |||
| BC073830.1 | - | 8 | 1164 | |||
| BC120993.2 | - | 6 | 960 | |||

C8orf31
Genomic Map