siRNA Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173829.3 | NP_776190.1 | 5 | 2273 | ||
| XM_011543344.2 | XP_011541646.1 | 4 | 2405 | |||
| GenBank mRNA | AK094073.1 | - | 2273 | |||
| AL832348.1 | - | 5 | 2230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SREK1IP1| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173829.3 | NP_776190.1 | 5 | 2273 | ||
| XM_011543344.2 | XP_011541646.1 | 4 | 2405 | |||
| GenBank mRNA | AK094073.1 | - | 2273 | |||
| AL832348.1 | - | 5 | 2230 | |||