siRNA Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174889.4 | NP_777549.1 | 1 | 213 | ||
| GenBank mRNA | AB183433.1 | - | 1 | 86 | ||
| AF087990.1 | - | 1 | 182 | |||
| AK026051.1 | - | 1 | 213 | |||
| AK291296.1 | - | 1 | 176 | |||
| BC001753.1 | - | 1 | 135 | 3354845 | ||
| BC022869.1 | - | 1 | 114 | |||
| BC033965.1 | - | 1 | 129 | 5288040 | ||
| BC070357.1 | - | 1 | 135 | |||
| KU178834.1 | - | 1 | 86 | |||
| KU178835.1 | - | 1 | 86 | |||
| KU178836.1 | - | 1 | 86 | |||

NDUFAF2
Genomic Map