siRNA Details
| Gene Symbol: FAM84B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174911.4 | NP_777571.1 | 2 | 1662 | ||
| XM_017013107.1 | XP_016868596.1 | 2 | 1709 | |||
| XM_017013108.1 | XP_016868597.1 | 3 | 1641 | |||
| GenBank mRNA | AK292824.1 | - | 2 | 1587 | ||
| BC052957.1 | - | 2 | 1417 | 5262890 | ||

FAM84B
Genomic Map