siRNA Details
| Gene Symbol: STPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174952.2 | NP_777612.1 | 4 | 516 | ||
| XM_011531886.2 | XP_011530188.1 | 5 | 908 | |||
| XM_017008050.1 | XP_016863539.1 | 4 | 749 | |||
| GenBank mRNA | BC036870.1 | - | 4 | 516 | 5226859 | |

STPG2
Genomic Map