siRNA Details
| Gene Symbol: SVOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139456.1 | NP_001132928.1 | 9 | 821 | ||
| NM_174959.3 | NP_777619.1 | 5 | 438 | |||
| XM_005250143.3 | XP_005250200.1 | 10 | 1225 | |||
| XM_011515797.2 | XP_011514099.1 | 6 | 512 | |||
| XM_017011746.1 | XP_016867235.1 | 7 | 627 | |||
| XM_017011747.1 | XP_016867236.1 | 6 | 452 | |||
| XM_017011748.1 | XP_016867237.1 | 6 | 457 | |||
| GenBank mRNA | BC036796.1 | - | 5 | 438 | 5590416 | |
| BX648789.1 | - | 5 | 4058 | |||

SVOPL
Genomic Map