siRNA Details
| Gene Symbol: JAZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175061.3 | NP_778231.2 | 5 | 2227 | ||
| XM_006715656.1 | XP_006715719.1 | 6 | 2103 | |||
| XM_017011831.1 | XP_016867320.1 | 5 | 1995 | |||
| XM_017011832.1 | XP_016867321.1 | 5 | 1927 | |||
| GenBank mRNA | AK000711.1 | - | 2 | 1454 | ||
| AK091311.1 | - | 5 | 2152 | |||
| AL832491.1 | - | 3 | 1805 | |||
| AL834234.1 | - | 4 | 1946 | |||
| AY372319.1 | - | 5 | 2005 | |||
| BC042441.1 | - | 6 | 2016 | 4831063 | ||
| BC047229.1 | - | 5 | 2014 | |||

JAZF1
Genomic Map