siRNA Details
| Gene Symbol: TDRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175075.4 | NP_778250.2 | 4 | 1003 | ||
| XM_006716232.3 | XP_006716295.1 | 3 | 983 | |||
| XM_017013113.1 | XP_016868602.1 | 3 | 1317 | |||
| GenBank mRNA | AK126209.1 | - | 1 | 468 | ||
| AY194292.2 | - | 3 | 821 | |||
| BX648606.1 | - | 2 | 606 | |||

TDRP
Genomic Map