siRNA Details
Gene Symbol: SOWAHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175873.5 | NP_787069.4 | 1 | 2419 | ||
GenBank mRNA | AK090823.1 | - | 1 | 846 | ||
AK092782.1 | - | 1 | 1362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SOWAHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175873.5 | NP_787069.4 | 1 | 2419 | ||
GenBank mRNA | AK090823.1 | - | 1 | 846 | ||
AK092782.1 | - | 1 | 1362 |