siRNA Details
| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175884.4 | NP_787080.2 | 1 | 1537 | ||
| GenBank mRNA | AK093350.1 | - | 1 | 1233 | ||
| BC013906.2 | - | 2 | 1240 | 3637017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC71L| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175884.4 | NP_787080.2 | 1 | 1537 | ||
| GenBank mRNA | AK093350.1 | - | 1 | 1233 | ||
| BC013906.2 | - | 2 | 1240 | 3637017 | ||