siRNA Details
| Gene Symbol: C5orf51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175921.5 | NP_787117.3 | 6 | 5097 | ||
| XM_005248289.4 | XP_005248346.1 | 5 | 5013 | |||
| XM_011514032.2 | XP_011512334.1 | 7 | 5226 | |||
| GenBank mRNA | AK094002.1 | - | 1 | 1933 | ||
| AL833916.1 | - | 1 | 2199 | |||

C5orf51
Genomic Map