siRNA Details
| Gene Symbol: IFNE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176891.4 | NP_795372.1 | 1 | 1195 | ||
| GenBank mRNA | AY190045.1 | - | 1 | 582 | ||
| AY358570.1 | - | 1 | 1196 | |||
| BC100871.2 | - | 1 | 582 | 40003063 | ||
| BC100872.2 | - | 1 | 582 | 40003065 | ||
| BC100873.1 | - | 1 | 582 | 40003066 | ||

IFNE
Genomic Map