siRNA Details
| Gene Symbol: TGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099691.2 | NP_001093161.1 | 5 | 628 | ||
| NM_001308158.1 | NP_001295087.1 | 5 | 798 | |||
| NM_001308159.1 | NP_001295088.1 | 5 | 795 | |||
| NM_003236.3 | NP_003227.1 | 5 | 631 | |||
| GenBank mRNA | AF149096.1 | - | 5 | 388 | ||
| AF149097.1 | - | 5 | 385 | |||
| AF149098.1 | - | 5 | 385 | |||
| AK290151.1 | - | 5 | 442 | |||
| AK309066.1 | - | 5 | 798 | |||
| AK309220.1 | - | 5 | 647 | |||
| AK312899.1 | - | 5 | 439 | |||
| BC005308.1 | - | 5 | 439 | 3828239 | ||
| BT006833.1 | - | 5 | 380 | |||
| K03222.1 | - | 5 | 417 | |||
| M31172.1 | - | 5 | 417 | |||
| X70340.1 | - | 5 | 414 | |||

TGFA
Genomic Map