siRNA Details
| Gene Symbol: LCN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178536.3 | NP_848631.2 | 6 | 673 | ||
| XM_006717065.3 | XP_006717128.1 | 6 | 2273 | |||
| XM_011518560.2 | XP_011516862.1 | 6 | 2425 | |||
| XM_011518561.2 | XP_011516863.2 | 5 | 1965 | |||
| XM_011518562.2 | XP_011516864.1 | 6 | 1826 | |||
| XM_017014631.1 | XP_016870120.1 | 6 | 1496 | |||
| XM_017014632.1 | XP_016870121.1 | 6 | 1416 | |||
| GenBank mRNA | BC041168.1 | - | 5 | 1788 | 5165480 | |

LCN12
Genomic Map