siRNA Details
| Gene Symbol: ZNF680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178558.4 | NP_848653.2 | 4 | 2162 | ||
| XM_006715959.2 | XP_006716022.1 | 5 | 2438 | |||
| XM_017012126.1 | XP_016867615.1 | 7 | 2744 | |||
| XM_017012127.1 | XP_016867616.1 | 6 | 2556 | |||
| XM_017012128.1 | XP_016867617.1 | 6 | 2626 | |||
| XM_017012129.1 | XP_016867618.1 | 5 | 2288 | |||
| GenBank mRNA | AK074911.1 | - | 1 | 1442 | ||
| AK122937.1 | - | 4 | 2123 | |||
| BC030700.1 | - | 4 | 2146 | 4837196 | ||

ZNF680
Genomic Map