siRNA Details
| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178562.4 | NP_848657.1 | 4, 5 | 636 | ||
| XM_006715960.3 | XP_006716023.1 | 4, 5 | 464 | |||
| GenBank mRNA | AF276891.1 | - | 4, 5 | 371 | ||
| AY236849.1 | - | 4, 5 | 460 | |||
| BC044244.1 | - | 4, 5 | 600 | 5759132 | ||

TSPAN33
Genomic Map