siRNA Details
| Gene Symbol: RSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282863.1 | NP_001269792.1 | 5 | 1554 | ||
| NM_001317942.1 | NP_001304871.1 | 5 | 1276 | |||
| NM_178565.4 | NP_848660.3 | 6 | 1955 | |||
| XM_011517018.1 | XP_011515320.1 | 6 | 1971 | |||
| XM_011517019.1 | XP_011515321.1 | 6 | 1968 | |||
| XM_017013395.1 | XP_016868884.1 | 6 | 1941 | |||
| GenBank mRNA | AK123023.1 | - | 6 | 1918 | ||
| AK123027.1 | - | 4 | 1085 | |||
| AY358166.1 | - | 6 | 1608 | |||
| BC027938.1 | - | 5 | 1276 | 5201681 | ||
| BC036554.1 | - | 5 | 1555 | 5265957 | ||

RSPO2
Genomic Map