siRNA Details
| Gene Symbol: SLC9B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300754.1 | NP_001287683.1 | 3 | 435 | ||
| NM_001300756.1 | NP_001287685.1 | 3 | 801 | |||
| NM_178833.5 | NP_849155.2 | 3 | 801 | |||
| XM_006714085.2 | XP_006714148.1 | 3 | 800 | |||
| XM_006714086.3 | XP_006714149.1 | 3 | 800 | |||
| GenBank mRNA | AK172823.1 | - | 3 | 801 | ||
| AK297010.1 | - | 3 | 346 | |||
| AK298506.1 | - | 2 | 118 | |||
| AK299888.1 | - | 3 | 360 | |||
| BC047447.1 | - | 3 | 331 | 5295722 | ||
| DC297189.1 | - | 3 | 436 | |||

SLC9B2
Genomic Map