siRNA Details
| Gene Symbol: LYRM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293735.1 | NP_001280664.1 | 4 | 633 | ||
| NM_181705.3 | NP_859056.2 | 5 | 715 | |||
| NR_121658.1 | 3 | 562 | ||||
| GenBank mRNA | AK000803.1 | - | 5 | 686 | ||
| BC037930.1 | - | 5 | 667 | 5284728 | ||
| BC047079.1 | - | 5 | 645 | 5286889 | ||
| BG714193.1 | - | 4 | 574 | |||

LYRM7
Genomic Map