siRNA Details
| Gene Symbol: LYRM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293735.1 | NP_001280664.1 | 4 | 967 | ||
| NM_181705.3 | NP_859056.2 | 5 | 1049 | |||
| NR_121658.1 | 3 | 896 | ||||
| GenBank mRNA | AK000803.1 | - | 5 | 1020 | ||
| BC037930.1 | - | 5 | 1001 | 5284728 | ||
| BC047079.1 | - | 5 | 979 | 5286889 | ||

LYRM7
Genomic Map